Learn More
Biomatik Corporation Human Carcinoembryonic Antigen-Related Cell Adhesion Molecule 8 (Ceacam8) Elisa Kit
Supplier:  Biomatik Corporation EKC33008
Encompass_Preferred
Human Carcinoembryonic Antigen-Related Cell Adhesion Molecule 8 (Ceacam8) Elisa Kit for detecting Carcinoembryonic Antigen-Related Cell Adhesion Molecule 8 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.156ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P31997; CEACAM8. Target Synonyms: Carcinoembryonic antigen CGM6; Carcinoembryonic antigen gene family member 6; Carcinoembryonic antigen related cell adhesion molecule 8; Carcinoembryonic antigen-related cell adhesion molecule 8; CD 66b; CD 67; CD66b; CD66b antigen; CD67; CD67 antigen; CEACAM 8; CEACAM8; CEAM8_HUMAN; CGM 6; CGM6; NCA 95; NCA95; Non-specific cross-reacting antigen NCA-95; Nonspecific cross reacting antigen NCA 95; Nonspecific cross reacting antigen NCA95. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3846