Learn More
Biomatik Corporation Human Cathepsin H (Ctsh) Elisa Kit
Supplier:  Biomatik Corporation EKC33045
Encompass_Preferred
Human Cathepsin H (Ctsh) Elisa Kit for detecting Cathepsin H in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P09668; CTSH. Target Synonyms: ACC 4; ACC 5; ACC4; ACC5; aleurain; CATH_HUMAN; cathepsin B3; cathepsin BA; Cathepsin H heavy chain; Cathepsin H light chain; Cathepsin H mini chain; CPSB; CTSH; minichain; N benzoylarginine beta naphthylamide hydrolase; N-benzoylarginine-beta-naphthylamide hydrolase; pro cathepsin H. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3854