Learn More
Biomatik Corporation Human Cc-Chemokine Receptor 2, Ccr2 Elisa Kit
Supplier:  Biomatik Corporation EKC33062
Encompass_Preferred
Human Cc-Chemokine Receptor 2, Ccr2 Elisa Kit for detecting Chemokine (C-C Motif) Receptor 2 in serum, plasma, tissue homogenates, cerebrospinal fluid (csf). Species Reactivity: Human (Homo sapiens). Detection Range: 23.44-1500pg/ml. Sensitivity: 5.86pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P41597; CCR2. Target Synonyms: CCR2; CMKBR2; C-C chemokine receptor type 2; C-C CKR-2; CC-CKR-2; CCR-2; CCR2; Monocyte chemoattractant protein 1 receptor; MCP-1-R; CD antigen CD192. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3856