Learn More
Biomatik Corporation Human Clip-Associating Protein 2 (Clasp2) Elisa Kit
Supplier:  Biomatik Corporation EKC33151
Encompass_Preferred
Human Clip-Associating Protein 2 (Clasp2) Elisa Kit for detecting Cytoplasmic Linker Associated Protein 2 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 23.5-1500pg/ml. Sensitivity: 5.86pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O75122; CLASP2. Target Synonyms: CLASP2; KIAA0627; CLIP-associating protein 2; Cytoplasmic linker-associated protein 2; Protein Orbit homolog 2; hOrbit2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3868