Learn More
Biomatik Corporation Human C-Myc Oncogene Product, C-Myc Elisa Kit
Supplier:  Biomatik Corporation EKC33173
Encompass_Preferred
Human C-Myc Oncogene Product, C-Myc Elisa Kit for detecting V-Myc Myelocytomatosis Viral Oncogene Homolog (Avian) in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.078ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P01106; MYC. Target Synonyms: AU016757; Avian myelocytomatosis viral oncogene homolog; bHLHe39; c Myc; Cellular myelocytomatosis oncogene; Class E basic helix-loop-helix protein 39; MGC105490; MRTL; Myc; Myc protein; Myc proto oncogene protein; Myc proto-oncogene protein; myc-related translation/localization regulatory factor; MYC_HUMAN; Myc2; myca; MYCC; Myelocytomatosis oncogene a; Myelocytomatosis oncogene; Niard; Nird; oncogene c-Myc; Oncogene Myc; OTTHUMP00000158589; OTTHUMP00000227763; Proto-oncogene c-Myc; Protooncogene homologous to myelocytomatosis virus. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3873