Learn More
Biomatik Corporation Human Corticosteroid Binding Globulin (Cbg) Elisa Kit
Supplier:  Biomatik Corporation EKC33294
Encompass_Preferred
Human Corticosteroid Binding Globulin (Cbg) Elisa Kit for detecting Serpin Peptidase Inhibitor, Clade A (Alpha-1 Antiproteinase, Antitrypsin), Member 6 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 20-8000pmol/ml. Sensitivity: 20pmol/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P08185; SERPINA6. Target Synonyms: CBG; CBG_HUMAN; corticosteroid binding globulin; Corticosteroid-binding globulin; serine (or cysteine) proteinase inhibitor clade A (alpha 1 antiproteinase antitrypsin) member 6; Serpin A6; serpin peptidase inhibitor clade A (alpha 1 antiproteinase antitrypsin) member 6; Serpina6; Transcortin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3896