Learn More
Biomatik Corporation Human Cystic Fibrosis Transmembrane Conductance Regulator (Cftr) Elisa Kit
Supplier:  Biomatik Corporation EKC33364
Encompass_Preferred
Human Cystic Fibrosis Transmembrane Conductance Regulator (Cftr) Elisa Kit for detecting Cystic Fibrosis Transmembrane Conductance Regulator (Atp-Binding Cassette Sub-Family C, Member 7) in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 28-1800pg/ml. Sensitivity: 7pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P13569; CFTR. Target Synonyms: ABC 35; ABC35; ABCC 7; ABCC7; ATP binding cassette sub family C member 7; ATP Binding Cassette Superfamily C Member 7; ATP binding cassette transporter sub family C member 7; ATP-binding cassette sub-family C member 7; cAMP dependent chloride channel; cAMP-dependent chloride channel; CF; CFTR; CFTR/MRP; CFTR_HUMAN; Channel conductance controlling ATPase; Channel conductance-controlling ATPase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3902