Learn More
Biomatik Corporation Human Cytochrome C-1 (Cyc1) Elisa Kit
Supplier:  Biomatik Corporation EKC33372
Encompass_Preferred
Human Cytochrome C-1 (Cyc1) Elisa Kit for detecting Cytochrome C-1 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P08574; CYC1. Target Synonyms: Complex III subunit 4; Complex III subunit IV; CY1_HUMAN; CYC1; Cytochrome b c1 complex subunit 4; Cytochrome b-c1 complex subunit 4; Cytochrome bc1 complex subunit 4; Cytochrome c 1; Cytochrome c-1; Cytochrome c1; heme protein; mitochondrial; Ubiquinol cytochrome c reductase complex cytochrome c1 subunit; Ubiquinol-cytochrome-c reductase complex cytochrome c1 subunit; UQCR4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3904