Learn More
Biomatik Corporation Human Cytochrome P450 3A4 (Cyp3A4) Elisa Kit
Supplier:  Biomatik Corporation EKC33381
Encompass_Preferred
Human Cytochrome P450 3A4 (Cyp3A4) Elisa Kit for detecting Cytochrome P450, Family 3, Subfamily A, Polypeptide 4 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 23.5-1500pg/ml. Sensitivity: 5.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P08684; CYP3A4. Target Synonyms: CYP3A4; CYP3A3; Cytochrome P450 3A4; 1; 4-cineole 2-exo-monooxygenase; 1; 8-cineole 2-exo-monooxygenase; Albendazole monooxygenase; sulfoxide-forming; Albendazole sulfoxidase; CYPIIIA3; CYPIIIA4; Cholesterol 25-hydroxylase; Cytochrome P450 3A3; Cytochrome P450 HLp; Cytochrome P450 NF-25; Cytochrome P450-PCN1; Nifedipine oxidase; Quinine 3-monooxygenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3908