Learn More
Biomatik Corporation Human Cytokeratin 18, Ck-18 Elisa Kit
Supplier:  Biomatik Corporation EKC33390
Encompass_Preferred
Human Cytokeratin 18, Ck-18 Elisa Kit for detecting Keratin 18 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-15ng/ml. Sensitivity: 0.25ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P05783. Target Synonyms: Cell proliferation inducing gene 46 protein; Cell proliferation inducing protein 46; Cell proliferation-inducing gene 46 protein; CK 18; CK-18; CK18; CYK 18; CYK18; Cytokeratin 18; Cytokeratin endo B; Cytokeratin-18; K 18; K18; K1C18_HUMAN; KA18; Keratin 18; Keratin 18; type I; Keratin D; keratin; type I cytoskeletal 18; Keratin-18; Krt18. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3911