Learn More
Biomatik Corporation Human Hyaluronoglucosaminidase 1 (HYAL1) ELISA Kit
Supplier:  Biomatik Corporation EKU08378
Encompass_Preferred
Human Hyaluronoglucosaminidase 1 (HYAL1) ELISA Kit for detecting HYAL1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.125ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q12794. Gene ID: 3373. Target Synonyms: LUCA1; NAT6; Hyaluronidase-1; Lung carcinoma protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3036