Learn More
Biomatik Corporation Human Neuron Derived Neurotrophic Factor (NDNF) ELISA Kit
Supplier:  Biomatik Corporation EKU08389
Encompass_Preferred
Human Neuron Derived Neurotrophic Factor (NDNF) ELISA Kit for detecting NDNF in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8TB73. Gene ID: 79625. Target Synonyms: C4orf31. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3038