Learn More
Biomatik Corporation Mouse Tumor Necrosis Factor Receptor 1 (TNFR1) ELISA Kit
Supplier:  Biomatik Corporation EKU08400
Encompass_Preferred
Mouse Tumor Necrosis Factor Receptor 1 (TNFR1) ELISA Kit for detecting TNFR1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P25118. Gene ID: 21937. Target Synonyms: CD120A; P55; TNFRSF1A; TBP1; FPF; TNF-R; TNF-R-I; TNF-R55; TNFAR; TNFR55; TNFR60; P55-R; P60; Tumor necrosis factor receptor 1; Tumor necrosis factor-binding protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3042