Learn More
Biomatik Corporation Human Organic Cation/Ergothioneine Transporter (OCTN1) ELISA Kit
Supplier:  Biomatik Corporation EKU08465
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 78-5,000pg/mL. Sensitivity: 30pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q9H015. Target Synonyms: SLC22A4; ETT; UT2H; Solute Carrier Family 22 Member 4; ET transporter. Restriction: For Research Use Only.
Catalog No. 50-229-3050