Learn More
Biomatik Corporation Alpha-Tocopherol (TCPa) CLIA Kit
Supplier:  Biomatik Corporation EKU08650
Encompass_Preferred
Alpha-Tocopherol (TCPa) CLIA Kit for detecting TCPa in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 781-200,000ng/ml. Sensitivity: 271ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Target Synonyms: VE; Vitamin E; α-TCP; α-Tocopherol. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3055