Learn More
Biomatik Corporation Dog C-Peptide (CP) ELISA Kit
Supplier:  Biomatik Corporation EKU08682
Encompass_Preferred
Dog C-Peptide (CP) ELISA Kit for detecting CP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 98.77-8,000pg/ml. Sensitivity: 37.55pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P01321. Gene ID: 483665. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3058