Learn More
Biomatik Corporation Rat Factor Related Apoptosis Ligand (FASL) ELISA Kit
Supplier:  Biomatik Corporation EKU08968
Encompass_Preferred
Rat Factor Related Apoptosis Ligand (FASL) ELISA Kit for detecting FASL in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 7.8-500pg/ml. Sensitivity: 3.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P36940. Gene ID: 25385. Target Synonyms: CD178; CD95L; CD95-L; FASLG; FASLG; APT1LG1; APT1-LG1; TNFSF6; Fas Antigen Ligand; TNF Superfamily Member 6; Tumor Necrosis Factor (ligand) Superfamily,Member 6. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3077