Learn More
Biomatik Corporation Mouse Lactoperoxidase (LPO) ELISA Kit
Supplier:  Biomatik Corporation EKU08995
Encompass_Preferred
Mouse Lactoperoxidase (LPO) ELISA Kit for detecting LPO in breast milk, tear, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q5SW46. Gene ID: 76113. Target Synonyms: SPO; Salivary peroxidase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3079