Learn More
Biomatik Corporation Mouse Podocin (PDCN) ELISA Kit
Supplier:  Biomatik Corporation EKU09037
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Mouse. Detection Range: 0.156-10ng/mL. Sensitivity: 0.054ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q91X05. Target Synonyms: NPHS2; NPHS-2; Nephrosis 2; Idiopathic Steroid-Resistant. Restriction: For Research Use Only.
Catalog No. 50-229-3084