Learn More
Biomatik Corporation Rat Lectin Galactoside Binding, Soluble 3 Binding Protein (LGALS3BP) ELISA Kit
Supplier:  Biomatik Corporation EKU09083
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenatesand other biological fluids. Species Reactivity: Rat. Detection Range: 1.56-100ng/mL. Sensitivity: 0.64ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: MAC-2-BP; LGALS3-BP; Galectin-3-Binding Protein; L3 Antigen; Mac-2-Binding Protein; Tumor-associated antigen 90K; Serum Protein 90K; Basement membrane autoantigen p105. Restriction: For Research Use Only.
Catalog No. 50-229-3091