Learn More
Biomatik Corporation Human Cytochrome C Oxidase Subunit I (COX1) ELISA Kit
Supplier:  Biomatik Corporation EKU09152
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.114ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: MT-CO1; MTCO1; COI; COXI; Mitochondrially Encoded Cytochrome C Oxidase I; Cytochrome c oxidase polypeptide I. Restriction: For Research Use Only.
Catalog No. 50-229-3096