Learn More
Biomatik Corporation Mouse Tumor Necrosis Factor Alpha Induced Protein 2 (TNFaIP2) ELISA Kit
Supplier:  Biomatik Corporation EKU09221
Encompass_Preferred
Mouse Tumor Necrosis Factor Alpha Induced Protein 2 (TNFaIP2) ELISA Kit for detecting TNFaIP2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.12-200pg/ml. Sensitivity: 1.15pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q61333. Gene ID: 21928. Target Synonyms: Primary response gene B94 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3103