Learn More
Biomatik Corporation Rat Cold Inducible RNA Binding Protein (CIRBP) ELISA Kit
Supplier:  Biomatik Corporation EKU09238
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Rat. Detection Range: 78-5,000pg/mL. Sensitivity: 32pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: CIRP; A18HNRNP; Glycine-Rich RNA Binding Protein. Restriction: For Research Use Only.
Catalog No. 50-229-3105