Learn More
Biomatik Corporation Mouse Autophagy Related Protein 12 (ATG12) ELISA Kit
Supplier:  Biomatik Corporation EKU09303
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Mouse. Detection Range: 31.2-2,000pg/mL. Sensitivity: 12.1pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: APG12; APG12L; HAPG12; Ubiquitin-like protein ATG12. Restriction: For Research Use Only.
Catalog No. 50-229-3112