Learn More
Biomatik Corporation Mouse Thrombospondin 1 (THBS1) CLIA Kit
Supplier:  Biomatik Corporation EKU09841
Encompass_Preferred
Mouse Thrombospondin 1 (THBS1) CLIA Kit for detecting THBS1 in plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 5.49-4,000pg/ml. Sensitivity: 1.99pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P35441. Target Synonyms: TSP1; Thrombospondin-1p180. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3145