Learn More
Biomatik Corporation Human Solute Carrier Family 6, Member 19 (SLC6A19) ELISA Kit
Supplier:  Biomatik Corporation EKU10225
Encompass_Preferred
Human Solute Carrier Family 6, Member 19 (SLC6A19) ELISA Kit for detecting SLC6A19 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.258ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q695T7. Gene ID: 340024. Target Synonyms: B0AT1; HND; Sodium-Dependent Neutral Amino Acid Transporter B (0) AT1; Hartnup Disease; System B (0) neutral amino acid transporter AT1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3167