Learn More
Biomatik Corporation Human Thyrotropin Releasing Hormone (TRH) ELISA Kit
Supplier:  Biomatik Corporation EKU10240
Encompass_Preferred
Human Thyrotropin Releasing Hormone (TRH) ELISA Kit for detecting TRH in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 7.8-500pg/ml. Sensitivity: 2.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P20396. Gene ID: 7200. Target Synonyms: TRF; Thyrotropin-Releasing Factor; Thyroliberin; Protirelin; Pro-tirelin; Prothyroliberin; TSH-releasing factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3171