Learn More
Biomatik Corporation Human Apolipoprotein B (APOB) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU10268
Encompass_Preferred
Human Apolipoprotein B (APOB) High-sensitive ELISA Kit for detecting APOB in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.72ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P04114. Gene ID: 338. Target Synonyms: Apo-B; FLDB; Apo B-100; Apo B-48. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3173