Learn More
Biomatik Corporation Rat Fibroblast Growth Factor 15 (FGF15) Wide-range ELISA Kit
Supplier:  Biomatik Corporation EKU10546
Encompass_Preferred
Rat Fibroblast Growth Factor 15 (FGF15) Wide-range ELISA Kit for detecting FGF15 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8VI81. Gene ID: 170582. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3185