Learn More
Biomatik Corporation Human Salusin Beta (SALb) Wide-range ELISA Kit
Supplier:  Biomatik Corporation EKU10573
Encompass_Preferred
Human Salusin Beta (SALb) Wide-range ELISA Kit for detecting SALb in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 24.69-2,000pg/ml. Sensitivity: 8.87pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8N2E6. Gene ID: 27433. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3191