Learn More
Biomatik Corporation Human Glutamate Receptor, Ionotropic, N-Methyl-D-Aspartate 2A (GRIN2A) Anti-antigen ELISA Kit
Supplier:  Biomatik Corporation EKU10639
Encompass_Preferred
Human Glutamate Receptor, Ionotropic, N-Methyl-D-Aspartate 2A (GRIN2A) Anti-antigen ELISA Kit for detecting GRIN2A in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.31ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h, 30min. Accession Number: Q12879. Gene ID: 2903. Target Synonyms: NMDAR2A; NR2A; Glutamate [NMDA] receptor subunit epsilon-1; N-methyl D-aspartate receptor subtype 2A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3198