Learn More
Biomatik Corporation Rat Aquaporin 1 (AQP1) ELISA Kit
Supplier:  Biomatik Corporation EKU10657
Encompass_Preferred
Rat Aquaporin 1 (AQP1) ELISA Kit for detecting AQP1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.11ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P29975. Gene ID: 25240. Target Synonyms: AQP-CHIP; CHIP28; CO; Channel-Forming Integral Protein,28kDa; Aquaporin-CHIP; Urine water channel; Water channel protein for red blood cells and kidney proximal tubule. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3201