Learn More
Biomatik Corporation Chicken Brain Natriuretic Peptide (BNP) ELISA Kit
Supplier:  Biomatik Corporation EKU10685
Encompass_Preferred
Chicken Brain Natriuretic Peptide (BNP) ELISA Kit for detecting BNP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 12.35-1,000pg/ml. Sensitivity: 4.64pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P18908. Gene ID: 395765. Target Synonyms: GC-B; B-Type Natriuretic Peptide; Ventricular Natriuretic Peptide; Gamma-brain natriuretic peptide; Brain natriuretic peptide 32. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3203