Learn More
Biomatik Corporation Mouse Coagulation Factor XIII A1 Polypeptide (F13A1) ELISA Kit
Supplier:  Biomatik Corporation EKU10731
Encompass_Preferred
Mouse Coagulation Factor XIII A1 Polypeptide (F13A1) ELISA Kit for detecting F13A1 in plasma. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000ng/ml. Sensitivity: 6.3ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8BH61. Gene ID: 74145. Target Synonyms: F13-A1; FXIIIA1; Protein-glutamine gamma-glutamyltransferase A chain; Transglutaminase A chain; Coagulation factor XIII A chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3211