Learn More
Biomatik Corporation Mouse Cytochrome C (CYCS) ELISA Kit
Supplier:  Biomatik Corporation EKU10778
Encompass_Preferred
Mouse Cytochrome C (CYCS) ELISA Kit for detecting CYCS in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.62ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P62897. Gene ID: 13063. Target Synonyms: Cyt-C; HCS; CYC; CytC. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3219