Learn More
Biomatik Corporation Mouse Dopachrome Tautomerase (DCT) ELISA Kit
Supplier:  Biomatik Corporation EKU10798
Encompass_Preferred
Mouse Dopachrome Tautomerase (DCT) ELISA Kit for detecting DCT in tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P29812. Gene ID: 13190. Target Synonyms: TYRP2; TRP-2; DT; Dopachrome Delta-Isomerase; Tyrosine-Related Protein 2; L-dopachrome Delta-isomerase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3221