Learn More
Biomatik Corporation Rat Fibroblast Growth Factor 2, Basic (FGF2) ELISA Kit
Supplier:  Biomatik Corporation EKU10828
Encompass_Preferred
Rat Fibroblast Growth Factor 2, Basic (FGF2) ELISA Kit for detecting FGF2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.6pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P13109. Gene ID: 54250. Target Synonyms: B-FGF; BFGF; FGFB; HBGH-2; Basic Fibroblast Growth Factor; Heparin-binding growth factor 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3225