Learn More
Biomatik Corporation Mouse Follicle Stimulating Hormone (FSH) CLIA Kit
Supplier:  Biomatik Corporation EKU10840
Encompass_Preferred
Mouse Follicle Stimulating Hormone (FSH) CLIA Kit for detecting FSH in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 703.1-180,000pg/ml. Sensitivity: 284.3pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Accession Number: P01216. Gene ID: 12640. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3229