Learn More
Biomatik Corporation Human Ubiquitin Carboxyl Terminal Hydrolase L1 (UCHL1) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU10971
Encompass_Preferred
Human Ubiquitin Carboxyl Terminal Hydrolase L1 (UCHL1) High-sensitive ELISA Kit for detecting UCHL1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P09936. Gene ID: 7345. Target Synonyms: PARK5; PGP9.5; Uch-L1; Neuron cytoplasmic protein 9.5; Ubiquitin thioesterase L1; Ubiquitin carboxyl-terminal hydrolase isozyme L1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3244