Learn More
Biomatik Corporation S-Adenosyl Methionine (SAM) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11249
Encompass_Preferred
S-Adenosyl Methionine (SAM) Mini sample ELISA Kit for detecting SAM in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Universal. Detection Range: 12.35-1,000ng/ml. Sensitivity: 5.51ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 3h. Target Synonyms: SAMe; SAM-e; S-Adenosylmethionine; S-(5'-Adenosyl)-L-Methionine Chloride. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3272