Learn More
Biomatik Corporation Human Monocyte Chemotactic Protein 4 (MCP4) ELISA Kit
Supplier:  Biomatik Corporation EKU11293
Encompass_Preferred
Human Monocyte Chemotactic Protein 4 (MCP4) ELISA Kit for detecting MCP4 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q99616. Gene ID: 6357. Target Synonyms: CCL13; SCYA13; NCC1; SCYL1; CKb10; Chemokine C-C-Motif Ligand 13; Small Inducible Cytokine Subfamily A (Cys-Cys) Member 13; CK-beta-10; Monocyte chemoattractant 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3276