Learn More
Biomatik Corporation Human Delta-Like Protein 4 (Dll4) Elisa Kit
Supplier:  Biomatik Corporation EKC33430
Encompass_Preferred
Human Delta-Like Protein 4 (Dll4) Elisa Kit for detecting Delta-Like 4 (Drosophila) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 23.5-1500pg/ml. Sensitivity: 5.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9NR61; DLL4. Target Synonyms: AOS6; Delta 4; delta 4 precursor; Delta ligand 4; delta ligand 4 precursor; Delta like 4; Delta like 4 homolog; Delta like 4 protein; Delta like canonical Notch ligand 4; Delta like protein 4; Delta-like 4 (Drosophila); Delta-like protein 4; Delta4; DLL 4; Dll4; DLL4_HUMAN; Drosophila Delta homolog 4; hdelta2; Homeobox protein DLL-4; MGC126344; Notch ligand delta 2; Notch ligand DLL4; Notch ligand DLL4 precursor; XDLL-4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3921