Learn More
Biomatik Corporation Human Desmogleins 1, Dsg-1 Elisa Kit
Supplier:  Biomatik Corporation EKC33446
Encompass_Preferred
Human Desmogleins 1, Dsg-1 Elisa Kit for detecting Desmoglein 1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 23.5-1500pg/ml. Sensitivity: 5.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q02413; DSG1. Target Synonyms: DSG1; CDHF4; Desmoglein-1; Cadherin family member 4; Desmosomal glycoprotein 1; DG1; DGI; Pemphigus foliaceus antigen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3926