Learn More
Biomatik Corporation Human E3 Ubiquitin-Protein Ligase Mylip (Mylip) Elisa Kit
Supplier:  Biomatik Corporation EKC33521
Encompass_Preferred
Human E3 Ubiquitin-Protein Ligase Mylip (Mylip) Elisa Kit for detecting Myosin Regulatory Light Chain Interacting Protein in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q8WY64; MYLIP. Target Synonyms: BM-023; BZF1; E3 ubiquitin protein ligase MYLIP; E3 ubiquitin-protein ligase MYLIP; Idol; Inducible degrader of the LDL-receptor; Lipoprotein receptor; MIR; MYLIP; MYLIP_HUMAN; Myosin regulatory light chain interacting protein; PP5242. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3934