Learn More
Biomatik Corporation Human Early Prostate Cancer Antigen 2 (Epca2) Elisa Kit
Supplier:  Biomatik Corporation EKC33530
Encompass_Preferred
Human Early Prostate Cancer Antigen 2 (Epca2) Elisa Kit for detecting Early Prostate Cancer Antigen 2 (Epca2) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 12.5-400ng/ml. Sensitivity: 6.25ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3936