Learn More
Biomatik Corporation Human calcitonin gene related peptide, CGRP ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC32941
Encompass_Preferred
The Human calcitonin gene related peptide, CGRP ELISA Kit allows for quantitative determination of calcitonin gene related peptide, CGRP in serum, plasma, cell culture supernates and tissue homogenates. Species Reactivity: Human. Detection Range: 1.56 pg/mL-100 pg/mL. Sensitivity: 0.39 pg/mL. Format: 96-Well Plate. Assay Type: ELISA. Detection Method: Colorimetric. Assay Time: 1-5 hours. Restriction: For Research Use Only.
Catalog No. 50-149-7733