Learn More
Biomatik Corporation Human Endothelial Lipase, El Elisa Kit
Supplier:  Biomatik Corporation EKC33568
Encompass_Preferred
Human Endothelial Lipase, El Elisa Kit for detecting Lipase, Endothelial in serum, plasma, cell culture supernates, tissue homogenates, urine. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.156ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9Y5X9; LIPG. Target Synonyms: EDL; EL; Endothelial cell-derived lipase; Endothelial lipase; Endothelial lipase precursor; Lipase endothelial; LIPE_HUMAN; LIPG; Lipoprotein lipase H; PRO719. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3944