Learn More
Biomatik Corporation Human Fibulin-1 (Fbln1) Elisa Kit
Supplier:  Biomatik Corporation EKC33718
Encompass_Preferred
Human Fibulin-1 (Fbln1) Elisa Kit for detecting Fibulin-1(Fbln1) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 6.25-400 ug/ml. Sensitivity: 1.56ug/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P23142; FBLN1. Target Synonyms: Basement membrane protein 90; BM 90; FBLN; Fbln1; FBLN1_HUMAN; FIBL 1; FIBL-1; FIBL1; Fibulin 1; Fibulin-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3967