Learn More
Biomatik Corporation Human Fms-Like Tyrosine Kinase 3, Flt3 Elisa Kit
Supplier:  Biomatik Corporation EKC33734
Encompass_Preferred
Human Fms-Like Tyrosine Kinase 3, Flt3 Elisa Kit for detecting Fms-Related Tyrosine Kinase 3 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P36888; FLT3. Target Synonyms: CD 135; CD135; CD135 antigen; Fetal liver kinase 2; FL cytokine receptor; Flk 2; Flk2; Flt 3; FLT-3; Flt3; FLT3_HUMAN; FMS like tyrosine kinase 3; Fms related tyrosine kinase 3; Fms-like tyrosine kinase 3; Growth factor receptor tyrosine kinase type III; Ly-72; OTTHUMP0000004234; Receptor type tyrosine protein kinase FLT3; Stem cell tyrosine kinase 1; Stk 1; STK-1; Stk1; Tyrosine protein kinase receptor FLT3; Tyrosine-protein kinase receptor FLT3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3973