Learn More
Biomatik Corporation Human Free Testosterone,F-TESTO ELISA Kit
Supplier:  Biomatik Corporation EKC33757
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Homo sapiens (Human). Detection Range: 7.5 pg/mL-1500 pg/mL. Sensitivity: 3.75 pg/mL. Format: 96-Well Plate. Assay Type: Competitive. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-3979